Learn More
AdipoGen DL-3-Hydroxyisobutyric acid Na
SDP
Catalog No. 502560751
Encompass_Preferred
Chemical. CAS 1219589-99-7. Formula C4H7NaO3. MW 126.09. Synthetic Intermediate formed in the valine catabolism. It is a reactant of the enzymes involved in branched chain amino acid metabolism 3-hydroxyisobutyrate dehydrogenase and 3-hydroxyisobutyryl-CoA hydrolase and it was found to have increased concentrations in MS patient metabolic profiles. This compound may be used to study the distribution, characterization and kinetics of these enzymes. Shown to inhibit key enzymes of the energy metabolism.
Catalog No. 50-256-0751 Supplier AdipoGen Supplier No. CDXH0085G001
Chemical. CAS 1219589-99-7. Formula C4H7NaO3. MW 126.09. Synthetic Intermediate formed in the valine catabolism. It is a reactant of the enzymes involved in branched chain amino acid metabolism 3-hydroxyisobutyrate dehydrogenase and 3-hydroxyisobutyryl-CoA hydrolase and it was found to have increased concentrations in MS patient metabolic profiles. This compound may be used to study the distribution, characterization and kinetics of these enzymes. Shown to inhibit key enzymes of the energy metabolism.