Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-3-Methyl-2-butanol, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
598-75-4
C5H12O
88.15
MFCD00004527
MXLMTQWGSQIYOW-UHFFFAOYSA-N
3-methyl-2-butanol,sec-isoamyl alcohol,2-butanol, 3-methyl,2-methyl-3-butanol,isopropylmethylcarbinol,methylisopropylcarbinol,1,2-dimethylpropanol,1,2-dimethyl-1-propanol,dl-3-methyl-2-butanol,3-methyl-butan-2-ol
11732
CHEBI:77517
3-methylbutan-2-ol
CC(C)C(C)O
598-75-4
C5H12O
88.15
MFCD00004527
MXLMTQWGSQIYOW-UHFFFAOYSA-N
3-methyl-2-butanol,sec-isoamyl alcohol,2-butanol, 3-methyl,2-methyl-3-butanol,isopropylmethylcarbinol,methylisopropylcarbinol,1,2-dimethylpropanol,1,2-dimethyl-1-propanol,dl-3-methyl-2-butanol,3-methyl-butan-2-ol
11732
CHEBI:77517
3-methylbutan-2-ol
CC(C)C(C)O
Viscosity 3.8 mPa.s (25°C)
Flash Point 39°C
Density 0.8100g/mL
Quantity 25mL
Boiling Point 112.0°C
Refractive Index 1.4080 to 1.4100
Packaging Glass bottle
Color Colorless
Infrared Spectrum Authentic
Specific Gravity 0.81
Assay Percent Range 98%
Linear Formula (CH3)2CHCH(OH)CH3
Merck Index 15, 6105
Solubility Information Solubility in water: 28g/L (30°C). Other solubilities: miscible with alcohol and ether
Formula Weight 88.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material DL-3-Methyl-2-butanol
Show More Show Less

GHS P Statement
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning