Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-3-Methyl-2-oxopentanoic Acid, Sodium Salt 98+ %, Thermo Scientific™

Catalog No. AC189700250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC189700250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC189700250 Supplier Thermo Scientific Chemicals Supplier No. 189700250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66872-74-0
Molecular Formula C6H9NaO3
Molecular Weight (g/mol) 152.13
MDL Number MFCD00002584
InChI Key SMDJDLCNOXJGKC-UHFFFAOYNA-M
Synonym sodium 3-methyl-2-oxopentanoate, sodium 3-methyl-2-oxovalerate, 3-methyl-2-oxopentanoic acid sodium salt, ketoisoleucine sodium salt, 3-methyl-2-oxovaleric acid sodium salt, dl-alpha-keto-beta-methylvaleric acid sodium salt, +/--3-methyl-2-oxovaleric acid sodium salt, sodium dl-3-methyl-2-oxopentanoate, pentanoic acid, 3-methyl-2-oxo-, sodium salt, sodium, a-keto-, a-methylvalerate
PubChem CID 2724058
IUPAC Name sodium 3-methyl-2-oxopentanoate
SMILES [Na+].CCC(C)C(=O)C([O-])=O

Specifications

Linear Formula C2H5CH(CH3)COCO2Na
Quantity 25 g
Beilstein 03, III, 1237
Formula Weight 152.13
Chemical Name or Material DL-3-Methyl-2-oxopentanoic acid, sodium salt, 98+ %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.