Learn More
Medchemexpress LLC DL-3-Phenylalanine-13C-1 | 64193-01-7 | 98.03% | 50 MG
SDP
Catalog No. 5000412569
Encompass_Preferred
DL-3-Phenylalanine-13C-1 is the 13C labeled DL-3-Phenylalanine. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12569 Supplier Medchemexpress LLC Supplier No. HYB1732S350MG
DL-3-Phenylalanine-13C-1 is the 13C labeled DL-3-Phenylalanine. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.