Learn More
Medchemexpress LLC HY-W017162 100mg Medchemexpress, DL-3-Phenyllactic acid CAS:828-01-3 Purity:>98%
SDP
Catalog No. 501934628
Encompass_Preferred
Medchemexpress, HY-W017162 100mg DL-3-Phenyllactic acid CAS:828-01-3 DL-3-Phenyllactic acid is a broad-spectrum antimicrobial compound. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4628 Supplier Medchemexpress LLC Supplier No. HYW017162100MG
Medchemexpress, HY-W017162 100mg DL-3-Phenyllactic acid CAS:828-01-3 DL-3-Phenyllactic acid is a broad-spectrum antimicrobial compound. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.