Learn More
Medchemexpress LLC DL-3-phenyllactic acid | 828-01-3 | MFCD00065928 | 99.9% | 166.17 g/mol | C9H10O3 | 10 MG
SDP
Catalog No. 5000301487
Encompass_Preferred
DL-3-phenyllactic acid is a racemic analytical standard used as an endogenous metabolite reference in research and method development. It is a small organic acid (C9H10O3, CAS 828-01-3) with a molecular weight of about 166.17 g/mol, supplied as a high-purity solid for analytical applications.
  • High purity for use as an analytical standard and calibration reference.
  • Suitable for chromatography and mass spectrometry method development.
  • Small solid quantity packaging for reference and trace-level analyses.
  • Well-characterized chemical identifiers aid database matching and reporting.
  • Stable under recommended storage for consistent analytical results.
Catalog No. 50-003-01487 Supplier Medchemexpress LLC Supplier No. HYW017162R10MG
DL-3-phenyllactic acid is a racemic analytical standard used as an endogenous metabolite reference in research and method development. It is a small organic acid (C9H10O3, CAS 828-01-3) with a molecular weight of about 166.17 g/mol, supplied as a high-purity solid for analytical applications.
  • High purity for use as an analytical standard and calibration reference.
  • Suitable for chromatography and mass spectrometry method development.
  • Small solid quantity packaging for reference and trace-level analyses.
  • Well-characterized chemical identifiers aid database matching and reporting.
  • Stable under recommended storage for consistent analytical results.