Learn More
Medchemexpress LLC DL-3-phenyllactic acid | 828-01-3 | MFCD00065928 | 99.9% | 166.18 | C9H10O3 | 100 MG
SDP
Catalog No. 5000300343
Encompass_Preferred
DL-3-phenyllactic acid is an analytical standard and naturally occurring organic acid with broad-spectrum antimicrobial activity. It is present in honey and lactic-acid-fermented foods and is used in analytical and research applications, including method development and studies of antimicrobial and feed additive properties.
  • High purity suitable for analytical applications.
  • Available as solid and solution pack sizes for experimental flexibility.
  • Compatible with LC, GC, and MS methods as a reference standard.
  • Demonstrates broad-spectrum activity against bacteria and fungi.
  • Useful for research into feed additive alternatives to antibiotics.
Catalog No. 50-003-00343 Supplier Medchemexpress LLC Supplier No. HYW017162R100MG
DL-3-phenyllactic acid is an analytical standard and naturally occurring organic acid with broad-spectrum antimicrobial activity. It is present in honey and lactic-acid-fermented foods and is used in analytical and research applications, including method development and studies of antimicrobial and feed additive properties.
  • High purity suitable for analytical applications.
  • Available as solid and solution pack sizes for experimental flexibility.
  • Compatible with LC, GC, and MS methods as a reference standard.
  • Demonstrates broad-spectrum activity against bacteria and fungi.
  • Useful for research into feed additive alternatives to antibiotics.