Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-4-Hydroxymandelic Acid 95%, Thermo Scientific™

Catalog No. AC173610010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC173610010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC173610010 Supplier Thermo Scientific Chemicals Supplier No. 173610010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7244-02-2
Molecular Formula C6H8O4S2
Molecular Weight (g/mol) 208.25
InChI Key CCQMORVULNZXIA-UHFFFAOYSA-L
Synonym ethylenedithio diacetic acid, 1,2-bis carboxymethylthio ethane, 2-2-carboxymethylsulfanyl ethylsulfanyl acetic acid, 2-2-carboxymethylthio ethylthio acetic acid, 2-carboxymethyl sulfanyl ethyl sulfanyl acetic acid, ethylenedithiodiacetic acid, acmc-2097kv, ethylenebisthio diacetic acid, 1,2-ethylenebis thioglycolic acid, 2,2'-ethylenedithio diacetic acid
PubChem CID 310815
IUPAC Name 2-[2-(carboxymethylsulfanyl)ethylsulfanyl]acetic acid
SMILES C(CSCC(=O)O)SCC(=O)O

Specifications

Color White
Infrared Spectrum Authentic
Linear Formula HOC6H4CH(OH)CO2H
Quantity 1 g
Beilstein 10, 410
Formula Weight 168.15
Percent Purity ≥94%
Physical Form Crystalline Powder
Chemical Name or Material DL-4-Hydroxymandelic acid, 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.