Learn More
Cayman Chemical DL-4662 (hydrochloride), Purity: ≥98%, 50 mg
SDP
Catalog No. 502892939
Encompass_Preferred
A 3,4-dimethoxy derivative of the substituted cathinones: pentedrone and α-PVP that also features an ethylamine in place of the amino group that is typically attached to the alpha carbon of pentedrone; intended for forensic and research purposes
Catalog No. 50-289-2939 Supplier Cayman Chemical Supplier No. 900207750 MG
A 3,4-dimethoxy derivative of the substituted cathinones: pentedrone and α-PVP that also features an ethylamine in place of the amino group that is typically attached to the alpha carbon of pentedrone; intended for forensic and research purposes