Learn More
Bellco Glass, Inc. Delong 4 Baffle Shake Flask,1L 4 x-DP Bottom Baffles
SDP
Catalog No. 501945548
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 250mlDimensions: 128x235mmClosure Size: 38Case Quantity: 6
Catalog No. 50-194-5548 Supplier Bellco Glass, Inc. Supplier No. 254701000
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 250mlDimensions: 128x235mmClosure Size: 38Case Quantity: 6