Learn More
Medchemexpress LLC DL-5-hydroxylysine hydrochloride | 13204-98-3 | MFCD00012878 | 97.3% | 198.65 g/mol | C6H15ClN2O3 | 5 MG
SDP
Catalog No. 5000302248
Encompass_Preferred
DL-5-hydroxylysine hydrochloride (CAS 13204-98-3) is an analytical reference standard of 5-hydroxylysine, a post-translationally modified amino acid primarily found in collagen. Provided as a solid standard for analytical, calibration, and quality-control use in biochemical and proteomic assays.
  • Analytical standard for quantification and method validation.
  • Suitable as a calibration standard in LC-MS and HPLC assays.
  • High assay purity appropriate for reference and QC applications.
  • Solid form, stable when stored refrigerated at 4 °C.
  • Small pack size suitable for low-volume laboratory use.
Catalog No. 50-003-02248 Supplier Medchemexpress LLC Supplier No. HYW014930R5MG
DL-5-hydroxylysine hydrochloride (CAS 13204-98-3) is an analytical reference standard of 5-hydroxylysine, a post-translationally modified amino acid primarily found in collagen. Provided as a solid standard for analytical, calibration, and quality-control use in biochemical and proteomic assays.
  • Analytical standard for quantification and method validation.
  • Suitable as a calibration standard in LC-MS and HPLC assays.
  • High assay purity appropriate for reference and QC applications.
  • Solid form, stable when stored refrigerated at 4 °C.
  • Small pack size suitable for low-volume laboratory use.