Learn More
Cayman Chemical DL-a-Difluoromethylornithine
SDP
Catalog No. 502965628
Encompass_Preferred
An irreversible inhibitor of ornithine decarboxylase that suppresses polyamine biosynthesis; displays antiangiogenic, cytostatic, and antiparasitic effects
Catalog No. 50-296-5628 Supplier Cayman Chemical Supplier No. 16889500 MG
An irreversible inhibitor of ornithine decarboxylase that suppresses polyamine biosynthesis; displays antiangiogenic, cytostatic, and antiparasitic effects