Learn More
Enzo Life Sciences DL-alpha-Difluoromethylornithine. hydrochloride. monohydrate (10mg). CAS: 96020-91-6
SDP
Supplier:  Enzo Life Sciences ALX270283M010
Encompass_Preferred
Specific, irreversible inhibitor of ornithine decarboxylase (ODC), the rate-limiting enzyme in polyamine biosynthesis. Potent chemopreventive agent. Induces apoptosis. Arginase activity inhibitor. Alternative name: DFMO . HCl . H2O, Eflornithine . HCl . H2O, RMI-71782. Formula: C6H12F2N2O2 . HCl . H2O. MW: 236.6. Purity: ≥98% (TLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8460