Learn More
Enzo Life Sciences DL-alpha-Difluoromethylornithine. hydrochloride. monohydrate (50mg). CAS: 96020-91-6
SDP
Supplier:  Enzo Life Sciences ALX270283M050
Encompass_Preferred
Inhibitor of ornithine decarboxylase and arginase. Specific, irreversible inhibitor of ornithine decarboxylase (ODC), the rate-limiting enzyme in polyamine biosynthesis. Potent chemopreventive agent. Induces apoptosis. Arginase activity inhibitor. Alternative name: DFMO . HCl . H2O, Eflornithine . HCl . H2O, RMI-71782. Formula: C6H12F2N2O2 . HCl . H2O. MW: 236.6. MI: 14: 3522. Purity: ≥98% (TLC). Appearance: White to off-white solid. Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C. Handling: Hygroscopic.
Catalog No. 50-200-8461