Learn More
Enzo Life Sciences DL-alpha-Tocopherol (high purity) (1g). CAS: 10191-41-0
SDP
Supplier:  Enzo Life Sciences ALX460018G001
Encompass_Preferred
Alternative name: Vitamin E. Purity: ≥97% (Assay). Appearance: Slightly yellow viscous oil. Solubility: Soluble in 100% ethanol, ether, chloroform, acetone or vegetable oils. Long Term Storage: +4°C.
Catalog No. 50-200-9287