Learn More
Apexbio Technology LLC DL-Adrenaline 329-65-7 5g
SDP
Catalog No. 5000091368
Encompass_Preferred
DL-Adrenaline is a non-selective agonist targeting alpha- and beta-adrenergic receptors It is designed to activate these adrenergic receptors thereby modulating downstream cellular signaling pathways involved in vascular contraction cardiac stimulation and bronchodilation DL-Adrenaline exerts its biological activity primarily through adrenergic receptor activation Based on these pharmacological properties DL-Adrenaline holds research potential in the study of adrenergic receptor function and downstream signaling in cardiovascular respiratory and neurophysiological processes as well as serving as a reference molecule for experimental assays investigating receptor affinity selectivity and activation pathways
Catalog No. 50-000-91368 Supplier Apexbio Technology LLC Supplier No. B13265000
DL-Adrenaline is a non-selective agonist targeting alpha- and beta-adrenergic receptors It is designed to activate these adrenergic receptors thereby modulating downstream cellular signaling pathways involved in vascular contraction cardiac stimulation and bronchodilation DL-Adrenaline exerts its biological activity primarily through adrenergic receptor activation Based on these pharmacological properties DL-Adrenaline holds research potential in the study of adrenergic receptor function and downstream signaling in cardiovascular respiratory and neurophysiological processes as well as serving as a reference molecule for experimental assays investigating receptor affinity selectivity and activation pathways