Learn More
Medchemexpress LLC DL-Alanine-d3 | 53795-94-1 | 100 MG
SDP
Catalog No. 5000420682
Encompass_Preferred
DL-Alanine-d3 is the deuterium labeled DL-Alanine. It is a racemic compound of L- and D-alanine, used as a reducing and capping agent with silver nitrate aqueous solutions for nanoparticle production. It can also be utilized for researching transition metal chelation, such as Cu(II), Zn(II), and Cd(II). DL-alanine functions as a sweetener, classified with glycine and sodium saccharin, and plays a key role in the glucose-alanine cycle between tissues and the liver.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-20682 Supplier Medchemexpress LLC Supplier No. HYN2362S2100MG
DL-Alanine-d3 is the deuterium labeled DL-Alanine. It is a racemic compound of L- and D-alanine, used as a reducing and capping agent with silver nitrate aqueous solutions for nanoparticle production. It can also be utilized for researching transition metal chelation, such as Cu(II), Zn(II), and Cd(II). DL-alanine functions as a sweetener, classified with glycine and sodium saccharin, and plays a key role in the glucose-alanine cycle between tissues and the liver.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may impact the pharmacokinetic and metabolic profiles of drugs.