Learn More
Medchemexpress LLC DL-Alanine-d3 | 53795-94-1 | 25 MG
SDP
Catalog No. 5000420664
Encompass_Preferred
DL-Alanine-d3 is the deuterium labeled DL-Alanine. DL-alanine, an amino acid, is the racemic compound of L- and D-alanine. It is used as a reducing and capping agent with silver nitrate aqueous solutions for nanoparticle production, and for research of transition metal chelation. DL-alanine is also a sweetener and plays a role in the glucose-alanine cycle between tissues and liver.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-20664 Supplier Medchemexpress LLC Supplier No. 25MGHYN2362S2
DL-Alanine-d3 is the deuterium labeled DL-Alanine. DL-alanine, an amino acid, is the racemic compound of L- and D-alanine. It is used as a reducing and capping agent with silver nitrate aqueous solutions for nanoparticle production, and for research of transition metal chelation. DL-alanine is also a sweetener and plays a role in the glucose-alanine cycle between tissues and liver.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.