Learn More
Medchemexpress LLC DL-Alanine-d3 | 53795-94-1 | 5 MG
SDP
Catalog No. 5000420399
Encompass_Preferred
DL-Alanine-d3 is a deuterium labeled DL-Alanine, an amino acid that functions as the racemic compound of L- and D-alanine. It is intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS. Deuteration, in this context, is noted for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Employed as a reducing and capping agent with silver nitrate aqueous solutions for nanoparticle production
  • Can be utilized for researching transition metals chelation, including Cu(II), Zn(II), and Cd(II)
  • Functions as a sweetener, classified alongside glycine and sodium saccharin
  • Plays a crucial role in the glucose-alanine cycle between tissues and the liver
Catalog No. 50-004-20399 Supplier Medchemexpress LLC Supplier No. HYN2362S25MG
DL-Alanine-d3 is a deuterium labeled DL-Alanine, an amino acid that functions as the racemic compound of L- and D-alanine. It is intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS. Deuteration, in this context, is noted for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Employed as a reducing and capping agent with silver nitrate aqueous solutions for nanoparticle production
  • Can be utilized for researching transition metals chelation, including Cu(II), Zn(II), and Cd(II)
  • Functions as a sweetener, classified alongside glycine and sodium saccharin
  • Plays a crucial role in the glucose-alanine cycle between tissues and the liver