Learn More
Medchemexpress LLC DL-alanine-d3 | 53795-94-1 | MFCD00084106 | 98.0% | 92.11 g/mol | C3H4D3NO2 | 10 MG
SDP
Catalog No. 5000289634
Encompass_Preferred
DL-Alanine-d3 is a deuterium-labeled form of the amino acid alanine provided as a solid for analytical and tracer applications. It is used as an internal standard and isotopic tracer in mass spectrometry, metabolomics, and method development where precise quantitation and tracking of alanine are required.
  • Deuterium-labeled analog for use as an internal standard.
  • High chemical and isotopic purity suitable for quantitative analysis.
  • Available in small, premeasured amounts for analytical workflows.
  • Suitable for mass spectrometry, metabolomics, and tracer studies.
  • Stable solid form for straightforward storage and handling.
Catalog No. 50-002-89634 Supplier Medchemexpress LLC Supplier No. HYN2362S210MG
Add to Cart
Add to Cart
DL-Alanine-d3 is a deuterium-labeled form of the amino acid alanine provided as a solid for analytical and tracer applications. It is used as an internal standard and isotopic tracer in mass spectrometry, metabolomics, and method development where precise quantitation and tracking of alanine are required.
  • Deuterium-labeled analog for use as an internal standard.
  • High chemical and isotopic purity suitable for quantitative analysis.
  • Available in small, premeasured amounts for analytical workflows.
  • Suitable for mass spectrometry, metabolomics, and tracer studies.
  • Stable solid form for straightforward storage and handling.