Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Alanine, FCC, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18602280 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
25 kg
100 kg
Packaging:
Amber Glass Bottle
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-280 100 g Amber Glass Bottle
18-602-282 25 kg Fiber Drum
18-602-283 100 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-280 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1044100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

A1044, 302-72-7, C3H7NO2

Spectrum™ Chemical DL-Alanine, FCC is used as a nutritional supplement and a flavoring agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 302-72-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064408
InChI Key QNAYBMKLOCPYGJ-UHFFFAOYNA-N
IUPAC Name 2-aminopropanoic acid
SMILES CC(N)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Lead (Pb) 5 mg/kg
Loss on Drying 0.3%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98.5 to 101.5%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.