Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich DL-Alaninol

Catalog No. 1921715G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-217-15G 5 g
19-217-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-217-15G Supplier Sigma Aldrich Supplier No. 1921715G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dl-alaninol has been used in the synthesis of:
  • N6-α-(i-hydroxypropyl) lysine
  • Diastereomers of dl-α-amino alcohols
  • Enantiopure and racemic samples of 2-methyl-n-tosylaziridine
  • (+/-)-3-(5-dimethylcarbamoyl-pent-1-enyl)-n-(2-hydroxy-1-methyl-ethyl)benzamidetandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf. 192171.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database. to learn more, visit sigma.com/metlin

Specifications

CAS 6168-72-5
Boiling Point 173°C to 176°C (lit.)
Molecular Formula C3H9NO
Linear Formula CH3CH(NH2)CH2OH
Refractive Index n20/D 1.450 (literature)
MDL Number MFCD00008085
Synonym (+/-)-2-Amino-1-propanol
Density 0.943 g/mL (at 25°C (literature))
Quantity 5 g
Molecular Weight (g/mol) 75.11
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.