Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Alanyl-DL-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. A02231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A02231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A02231G Supplier TCI America Supplier No. A02231G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1999-45-7
Molecular Formula C12H16N2O3
Molecular Weight (g/mol) 236.271
MDL Number MFCD00008074
InChI Key OMNVYXHOSHNURL-UHFFFAOYSA-N
Synonym H-DL-Ala-DL-Phe-OH
PubChem CID 2080
IUPAC Name 2-(2-aminopropanoylamino)-3-phenylpropanoic acid
SMILES CC(C(=O)NC(CC1=CC=CC=C1)C(=O)O)N

Specifications

Color White
Quantity 1 g
Formula Weight 236.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Alanyl-DL-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.