Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Allothreonine (contains Threonine) 98.0+%, TCI America™
SDP

Catalog No. A0214100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0214100MG 100 mg
A02141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0214100MG Supplier TCI America Supplier No. A0214100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 144-98-9
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064268,MFCD00063722,MFCD00066665,MFCD00004526,MFCD00067248,MFCD00064269,MFCD00063722
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
Synonym l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonine
PubChem CID 99289
ChEBI CHEBI:28718
IUPAC Name 2-amino-3-hydroxybutanoic acid
SMILES CC(O)C(N)C(O)=O

Specifications

Color White
Quantity 100 mg
Formula Weight 119.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Allothreonine (contains Threonine)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.