Learn More
Medchemexpress LLC DL-alpha-tocopherol | 10191-41-0 | MFCD00072051 | 99.1% | 430.71 g/mol | C29H50O2 | 50 MG
SDP
Catalog No. 5000300466
Encompass_Preferred
DL-alpha-tocopherol is the racemic (synthetic) form of vitamin E supplied as an analytical standard for research and laboratory applications. It functions as a fat-soluble antioxidant and is commonly used as a reference material in assays and studies of oxidative protection in biological systems.
  • Synthetic form of vitamin E (racemic)
  • Fat-soluble antioxidant for protection against lipid peroxidation
  • Provided as a 50 mg analytical standard suitable for research assays
  • High purity for accurate analytical and experimental results
  • Molecular formula C29H50O2; molecular weight 430.71 g/mol
Catalog No. 50-003-00466 Supplier Medchemexpress LLC Supplier No. HYW020044R50MG
DL-alpha-tocopherol is the racemic (synthetic) form of vitamin E supplied as an analytical standard for research and laboratory applications. It functions as a fat-soluble antioxidant and is commonly used as a reference material in assays and studies of oxidative protection in biological systems.
  • Synthetic form of vitamin E (racemic)
  • Fat-soluble antioxidant for protection against lipid peroxidation
  • Provided as a 50 mg analytical standard suitable for research assays
  • High purity for accurate analytical and experimental results
  • Molecular formula C29H50O2; molecular weight 430.71 g/mol