Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-alpha-tocopherol | 10191-41-0 | MFCD00072051 | 99.8% | 430.71 g·mol⁻¹ | C29H50O2 | 1 ML
SDP

Catalog No. 5000300464
Encompass_Preferred

DL-alpha-tocopherol is the racemic (±) form of vitamin E used as a lipid-soluble antioxidant in biological research. It scavenges free radicals and protects cell membranes from oxidative damage, and is commonly used in cell-based and biochemical assays to study oxidative stress and cytoprotection against UVB-induced injury.

  • Acts as a lipid-soluble antioxidant protecting membranes from oxidative damage.
  • Protects human skin fibroblasts against UVB-induced cytotoxicity.
  • Provided as a ready-to-use DMSO solution for cell and biochemical assays.
  • High purity suitable for research applications (≈99.8%).
  • Soluble in organic solvents such as DMSO and insoluble in water.
  • Molecular weight 430.71 g·mol⁻¹; CAS 10191-41-0.

Catalog No. 50-003-00464 Supplier Medchemexpress LLC Supplier No. HYW02004410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-alpha-tocopherol is the racemic (±) form of vitamin E used as a lipid-soluble antioxidant in biological research. It scavenges free radicals and protects cell membranes from oxidative damage, and is commonly used in cell-based and biochemical assays to study oxidative stress and cytoprotection against UVB-induced injury.

  • Acts as a lipid-soluble antioxidant protecting membranes from oxidative damage.
  • Protects human skin fibroblasts against UVB-induced cytotoxicity.
  • Provided as a ready-to-use DMSO solution for cell and biochemical assays.
  • High purity suitable for research applications (≈99.8%).
  • Soluble in organic solvents such as DMSO and insoluble in water.
  • Molecular weight 430.71 g·mol⁻¹; CAS 10191-41-0.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.