Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-alpha-Tocopherol 1g | 1309573-60-1 | 1 G
SDP

Catalog No. 5000228086
Encompass_Preferred

DL-alpha-tocopherol methoxypolyethylene glycol succinate is a polyethylene glycol succinate derivative of DL-alpha-tocopherol used as a solubilizer and surfactant in biological and pharmaceutical formulations. It is supplied as a solution and characterized for solubility, storage, and purity.

  • provides amphiphilic solubilization for hydrophobic compounds
  • forms clear aqueous solutions: water ≥ 50 mg/mL
  • soluble in PBS at 25 mg/mL with ultrasonic treatment and warming
  • high purity by HPLC (98.0%)
  • polymeric chemical formula (C2H4O)nC34H56O5; reported molecular weight approximately 605 g/mol
  • store as solution at -20 °C for up to 2 years

Catalog No. 50-002-28086 Supplier Medchemexpress LLC Supplier No. HY1143621G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-alpha-tocopherol methoxypolyethylene glycol succinate is a polyethylene glycol succinate derivative of DL-alpha-tocopherol used as a solubilizer and surfactant in biological and pharmaceutical formulations. It is supplied as a solution and characterized for solubility, storage, and purity.

  • provides amphiphilic solubilization for hydrophobic compounds
  • forms clear aqueous solutions: water ≥ 50 mg/mL
  • soluble in PBS at 25 mg/mL with ultrasonic treatment and warming
  • high purity by HPLC (98.0%)
  • polymeric chemical formula (C2H4O)nC34H56O5; reported molecular weight approximately 605 g/mol
  • store as solution at -20 °C for up to 2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.