Learn More
Medchemexpress LLC DL-alpha-Tocopherol 5g | 1309573-60-1 | 5 G
SDP
Catalog No. 5000229164
Encompass_Preferred
DL-alpha-tocopherol methoxypolyethylene glycol succinate is a vitamin E-derived amphiphilic surfactant and solubilizing agent supplied as an aqueous solution for research use; it is used to aid solubilization, improve reaction performance, and support stability testing of modified drugs.
  • Acts as an amphiphilic surfactant and solubilizer.
  • Improves aqueous solubility of lipophilic compounds.
  • Enhances aqueous cross-coupling reaction performance (Suzuki-Miyaura reported).
  • Used for stability testing of modified drugs in biological fluids.
  • Typically provided as an aqueous solution at about 20 mg/mL.
Catalog No. 50-002-29164 Supplier Medchemexpress LLC Supplier No. HY1143625G
DL-alpha-tocopherol methoxypolyethylene glycol succinate is a vitamin E-derived amphiphilic surfactant and solubilizing agent supplied as an aqueous solution for research use; it is used to aid solubilization, improve reaction performance, and support stability testing of modified drugs.
  • Acts as an amphiphilic surfactant and solubilizer.
  • Improves aqueous solubility of lipophilic compounds.
  • Enhances aqueous cross-coupling reaction performance (Suzuki-Miyaura reported).
  • Used for stability testing of modified drugs in biological fluids.
  • Typically provided as an aqueous solution at about 20 mg/mL.