Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-alpha-Tocopherol acetate, MilliporeSigma™ Supelco™

Catalog No. 111015479 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5479 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5479 Supplier MilliporeSigma Supelco Supplier No. 47786
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 7695-91-2
Density 0.953 g/mL (at 25°C)
Flash Point 225°C
Physical Form Neat
Linear Formula C31H52O3
MDL Number MFCD00072052
Boiling Point 224°C (0.3 mmHg)
Synonym Vitamin E acetate; O-Acetyl-alpha-tocopherol; DL-alpha-Tocopherol acetate; DL-alpha-Tocopheryl acetate
Type Analytical Standard
Quantity 100 mg
Formula Weight 472.74
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.