Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-alpha-Tocopherol acetate powder, MP Biomedicals™

Catalog No. ICN10055525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10055525 25 g
ICN10055580 100 g
ICN10055590 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10055525 Supplier MP Biomedicals, Inc Supplier No. 0210055525
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7695-91-2
Molecular Formula C31H52O3
Molecular Weight (g/mol) 472.754
MDL Number MFCD00072042
InChI Key ZAKOWWREFLAJOT-CEFNRUSXSA-N
Synonym vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ
PubChem CID 86472
ChEBI CHEBI:32321
IUPAC Name [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate
SMILES CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Density 0.96g/mL at 20°C
Assay Percent Range 250U/g
Quantity 25 g
Physical Form Powder
Chemical Name or Material DL-α-Tocopherol acetate, Cell Biology and Analysis
EINECSNumber 231-710-0
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.