Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-α-Aminocaprylic acid, 99%, Thermo Scientific™

Catalog No. AC203220050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC203220050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC203220050 Supplier Thermo Scientific Chemicals Supplier No. 203220050

Chemical Identifiers

CAS 644-90-6
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00008102
InChI Key AKVBCGQVQXPRLD-UHFFFAOYSA-N
Synonym dl-2-aminocaprylic acid, dl-2-aminooctanoic acid, 2-aminocaprylic acid, octanoic acid, 2-amino, 2-amino-dl-caprylic acid, +--2-aminooctanoic acid, dl-alpha-aminocaprylic acid, 2-amino-octanoic acid, +/--2-aminooctanoic acid, caprylic acid, alpha-amino
PubChem CID 69522
ChEBI CHEBI:75145
IUPAC Name 2-aminooctanoic acid
SMILES CCCCCCC(C(=O)O)N

Specifications

Melting Point 260.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)5CH(NH2)COOH
Quantity 5 g
Beilstein 04, 461
Solubility Information Solubility in water: slightly soluble
Formula Weight 159.23
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material DL-α-Aminocaprylic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-424-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.