Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Aminoglutethimide, 99%

Catalog No. AAJ6218214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6218214 25 g
AAJ6218203 1 g
AAJ6218206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6218214 Supplier Thermo Scientific Chemicals Supplier No. J6218214
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 125-84-8 | C13H16N2O2 | 232.283 g/mol

Aminoglutethimide is used to decrease the production of sex hormones and suppress the growth of tumors that need sex hormones to grow. It blocks the production of steroids derived from cholesterol and is clinically used in the treatment of Cushing's syndrome and metastatic breast cancer. It is also a drug of abuse by body builders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (2 mg/ml at 20°C), methanol (50 mg/ml), ethanol (7 mg/ml at 25°C), DMSO (20 mg/ml at 25°C), and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125-84-8
Molecular Formula C13H16N2O2
Molecular Weight (g/mol) 232.283
MDL Number MFCD00010122
InChI Key ROBVIMPUHSLWNV-UHFFFAOYSA-N
Synonym aminoglutethimide, dl-aminoglutethimide, cytadren, orimeten, p-aminoglutethimide, elipten, 3-4-aminophenyl-3-ethylpiperidine-2,6-dione, aminoglutetimida, 3-4-aminophenyl-3-ethyl-2,6-piperidinedione, aminoglutethimidum
PubChem CID 2145
ChEBI CHEBI:2654
IUPAC Name 3-(4-aminophenyl)-3-ethylpiperidine-2,6-dione
SMILES CCC1(CCC(=O)NC1=O)C2=CC=C(C=C2)N

Specifications

Melting Point 152°C to 154°C
Color White
Quantity 25 g
Merck Index 14,440
Solubility Information Soluble in water (2mg/ml at 20°C),methanol (50mg/ml),ethanol (7mg/ml at 25°C),DMSO (20mg/ml at 25°C),and chloroform.
Formula Weight 232.28
Percent Purity 99%
Physical Form Powder
Chemical Name or Material DL-Aminoglutethimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-756-4
RTECSNumber MA4026950
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.