Learn More
Abcam DL-AP4 sodium salt (mM/ml), Glutamate antagonist, 100MM_ML
Catalog No. 502704526
Encompass_Preferred
MW 205.08 Da, Purity >99%. Broad spectrum glutamate receptor antagonist. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4526 Supplier Abcam Supplier No. AB146690100MM_ML
MW 205.08 Da, Purity >99%. Broad spectrum glutamate receptor antagonist. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement