Learn More
Apexbio Technology LLC DL-AP5(Synonyms: DL-2-Amino-5-phosphonopentanoic acid, DL-APV, AP5, DL-AP5), 10mg, CAS: 76326-31-3.
SDP
Catalog No. 5000060478
Encompass_Preferred
DL-AP5 (CAS 76326-31-3) is a competitive antagonist targeting the N-methyl-D-aspartate (NMDA) receptor a subtype of glutamate receptor critical for neuronal signaling in the central nervous system By competitively inhibiting glutamate binding at NMDA receptor sites DL-AP5 disrupts NMDA receptor-mediated signaling pathways thereby modulating synaptic transmission Due to this mechanism the compound is widely utilized in research examining neuronal communication processes and investigating pathophysiological mechanisms underlying neurological disorders
Catalog No. 50-000-60478 Supplier Apexbio Technology LLC Supplier No. B620410
DL-AP5 (CAS 76326-31-3) is a competitive antagonist targeting the N-methyl-D-aspartate (NMDA) receptor a subtype of glutamate receptor critical for neuronal signaling in the central nervous system By competitively inhibiting glutamate binding at NMDA receptor sites DL-AP5 disrupts NMDA receptor-mediated signaling pathways thereby modulating synaptic transmission Due to this mechanism the compound is widely utilized in research examining neuronal communication processes and investigating pathophysiological mechanisms underlying neurological disorders