Learn More
Enzo Life Sciences DL-AP5 (25mg). CAS: 76326-31-3
SDP
Supplier:  Enzo Life Sciences ALX550034M025
Encompass_Preferred
Potent and selective NMDA receptor antagonist. Alternative name: DL-2-Amino-5-phosphonopentanoic acid, DL-2-Amino-5-phosphonovaleric acid. Purity: ≥99% (HPLC, TLC). Solubility: Soluble in 100mM NaOH or 25mM water. Long Term Storage: Ambient.
Catalog No. 50-200-9461