Learn More
Abcam DL-AP5, NMDA glutamate site antagonist, 10MG
Catalog No. 502584725
Encompass_Preferred
MW 197.13 Da, Purity >99%. Competitive NMDA receptor glutamate site antagonist. Also available as the sodium salt (ab120271).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4725 Supplier Abcam Supplier No. AB12000410MG
MW 197.13 Da, Purity >99%. Competitive NMDA receptor glutamate site antagonist. Also available as the sodium salt (ab120271).
The product is subject to the following: Abcam Restricted Use Statement