Learn More
Selleck Chemical LLC DL-Arabinose
SDP
Catalog No. 5000003783
Encompass_Preferred
L-( )-Arabinose (DL-Arabinose Pectinose) is used as a substrate to identify differentiate and characterize pentose sugar isomerase(s)
Catalog No. 50-000-03783 Supplier Selleck Chemical LLC Supplier No. S634625MG
L-( )-Arabinose (DL-Arabinose Pectinose) is used as a substrate to identify differentiate and characterize pentose sugar isomerase(s)