Learn More
Medchemexpress LLC DL-arginine-13C6,15N4 hydrochloride | 95.0% | 220.59 | 13C6H15ClN4O2 (13C6,15N4 labeled) | 5 MG
SDP
Catalog No. 5000302981
Encompass_Preferred
DL-Arginine-13C6,15N4 hydrochloride is a stable isotope-labeled racemic arginine hydrochloride supplied for research applications. With six 13C and four 15N labeled atoms, it serves as an internal standard for mass spectrometry, metabolic tracing, and quantitative proteomics. Supplied as a high-purity solid in small packaging for analytical workflows and method development.
  • Contains six 13C and four 15N isotopic labels.
  • Suitable as an internal standard for mass spectrometry.
  • Useful for metabolic tracing and quantitative proteomics.
  • Supplied as a small-scale, high-purity solid for analytical use.
  • For research use only; not for human or clinical applications.
Catalog No. 50-003-02981 Supplier Medchemexpress LLC Supplier No. HYW014375S45MG
DL-Arginine-13C6,15N4 hydrochloride is a stable isotope-labeled racemic arginine hydrochloride supplied for research applications. With six 13C and four 15N labeled atoms, it serves as an internal standard for mass spectrometry, metabolic tracing, and quantitative proteomics. Supplied as a high-purity solid in small packaging for analytical workflows and method development.
  • Contains six 13C and four 15N isotopic labels.
  • Suitable as an internal standard for mass spectrometry.
  • Useful for metabolic tracing and quantitative proteomics.
  • Supplied as a small-scale, high-purity solid for analytical use.
  • For research use only; not for human or clinical applications.