Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dl-asparagine | 3130-87-8 | MFCD00064400 | 98.0% | 132.12 | C4H8N2O3 | 100 G
SDP

Catalog No. 5000300480
Encompass_Preferred

DL-Asparagine is the racemic mixture of the L- and D-enantiomers of the amino acid asparagine. It is supplied as a solid research reagent for use as an endogenous metabolite, in culture media, and as an analytical standard for laboratory applications.

  • Racemic mixture of L- and D-enantiomers.
  • High purity (98.0%) suitable for research applications.
  • Molecular weight 132.12; chemical formula C4H8N2O3.
  • Soluble in water for preparation of stock solutions.
  • Available in 100 g pack size for laboratory use.

Catalog No. 50-003-00480 Supplier Medchemexpress LLC Supplier No. HYW017442100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-Asparagine is the racemic mixture of the L- and D-enantiomers of the amino acid asparagine. It is supplied as a solid research reagent for use as an endogenous metabolite, in culture media, and as an analytical standard for laboratory applications.

  • Racemic mixture of L- and D-enantiomers.
  • High purity (98.0%) suitable for research applications.
  • Molecular weight 132.12; chemical formula C4H8N2O3.
  • Soluble in water for preparation of stock solutions.
  • Available in 100 g pack size for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.