Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-aspartic acid | 617-45-8 | MFCD00063083 | 98.0% | 133.10 g/mol | C4H7NO4 | 25 G
SDP

Catalog No. 5000303087
Encompass_Preferred

DL-Aspartic acid is the racemic form of the amino acid aspartic acid provided as a research-grade biochemical reagent for analytical and life-science applications. It is used as an assay reagent, analytical standard, or building block in experiments that do not require enantiopure material.

  • Racemic amino acid suitable for biochemical assays and analytical applications.
  • CAS number 617-45-8; molecular formula C4H7NO4; molecular weight 133.10 g/mol.
  • Water solubility ~10 mg/mL with warming; limited solubility in DMSO.
  • Stable as a powder under refrigerated or frozen storage per supplier guidance.
  • Available in small analytical and gram-scale package sizes for laboratory use.

Catalog No. 50-003-03087 Supplier Medchemexpress LLC Supplier No. 25GHYW015824
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-Aspartic acid is the racemic form of the amino acid aspartic acid provided as a research-grade biochemical reagent for analytical and life-science applications. It is used as an assay reagent, analytical standard, or building block in experiments that do not require enantiopure material.

  • Racemic amino acid suitable for biochemical assays and analytical applications.
  • CAS number 617-45-8; molecular formula C4H7NO4; molecular weight 133.10 g/mol.
  • Water solubility ~10 mg/mL with warming; limited solubility in DMSO.
  • Stable as a powder under refrigerated or frozen storage per supplier guidance.
  • Available in small analytical and gram-scale package sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.