Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Aspartic acid, 99+%, MP Biomedicals™

Catalog No. ICN10080383 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10080383 250 g
ICN10080390 500 g
ICN10080391 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10080383 Supplier MP Biomedicals, Inc Supplier No. 0210080383
Add to Cart
Add to Cart

Chemical Identifiers

CAS 617-45-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00063083
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart
PubChem CID 424
ChEBI CHEBI:22660
IUPAC Name 2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

Assay Percent Range 99+%
Quantity 250 g
Formula Weight 133.1
Physical Form Crystalline
Chemical Name or Material DL-Aspartic acid
EINECSNumber 210-513-3
Recommended Storage Store at room temperature.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.