Learn More
Description
- CAS Number: 617-45-8
- Molecular Formula: C4H7NO4
- Molecular Weight: 133.1
- Purity: ≥99.0%
- Melting Point: >300 °C (Lit.)
- Solubility: Soluble in NaOH 100 mg/ mL
- pH: 3.0 (Lit.)
- Optical Rotation: + 25.5 ° (Lit.)
- Biochemical Physiological Actions: DL-Aspartic acid is a racemic mixture of the proteinogenic amino acid L-aspartate and the non-proteinogenic amino acid D-aspartate, used in studies on factors and conditions that enhance semen and sperm quality.
- Usage Statement: Research or further manufacturing use only, not for direct human use.
Specifications
Specifications
| Assay Percent Range | 99+% |
| Quantity | 1 kg |
| Formula Weight | 133.1 |
| Percent Purity | ≥99.0% |
| Grade | Research |
| Physical Form | Crystalline |
| Chemical Name or Material | DL-Aspartic acid |
Safety and Handling
| EINECSNumber | 210-513-3 |
| Recommended Storage | Store at room temperature. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.