Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Aspartic acid, 99+%

Catalog No. AC105030050
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105030050 5 g Glass Bottle
AC105032500 250 g Plastic Bottle
AC105030010 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC105030050 Supplier Thermo Scientific Chemicals Supplier No. 105030050
  • $35.15
    $28.65 / Each of 1

    Save $6.50

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 617-45-8 | C4H7NO4 | 133.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-45-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00063083
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart
PubChem CID 424
ChEBI CHEBI:22660
IUPAC Name 2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

Ammonium 500ppm max.
Color Yellow
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 0.3% max. (105°C, 3 hrs)
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula HO2CCH2CH(NH2)CO2H
Beilstein 04, 483
Specific Rotation −0.3° to +0.3° (20°C, 589nm) (c=10, 2 N HCl)
Sulfate 500ppm max.
Merck Index 10, 853
Transmittance 95% min. (c=10, 2 N HCl, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Quantity 5 g
Formula Weight 133.1
Percent Purity 99.0 to 101.0% (Acid)
Physical Form Crystalline Low Melting Mass
Chemical Name or Material DL-Aspartic acid, 99+%
Show More Show Less
EINECSNumber 210-513-3
RTECSNumber CI9097800
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.