Learn More
Medchemexpress LLC DL-Aspartic acid-d3 | 14341-75-4 | MFCD00144167 | 136.12 | 5 MG
SDP
Catalog No. 5000388663
Encompass_Preferred
DL-Aspartic acid-d3 is a deuterium labeled L-Aspartic acid. L-Aspartic acid is an amino acid capable of penetrating the blood-brain barrier. It is often utilized in the preparation of prodrugs designed to target colon and cecal tissues, and is commonly used in studies of inflammatory conditions.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-88663 Supplier Medchemexpress LLC Supplier No. HYN0666S25MG
DL-Aspartic acid-d3 is a deuterium labeled L-Aspartic acid. L-Aspartic acid is an amino acid capable of penetrating the blood-brain barrier. It is often utilized in the preparation of prodrugs designed to target colon and cecal tissues, and is commonly used in studies of inflammatory conditions.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS