Learn More
Cayman Chemical DL-Aspartic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 14341-75-4, 25 mg
SDP
Catalog No. 502910989
Encompass_Preferred
An internal standard for the quantification of aspartic acid by GC- or LC-MS
Catalog No. 50-291-0989 Supplier Cayman Chemical Supplier No. 3483625 MG
An internal standard for the quantification of aspartic acid by GC- or LC-MS