Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-alpha-Bromophenylacetic acid, 97%

Catalog No. AC107270050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107270050 5 g Glass bottle
AC107270250 25 g Glass bottle
AC107271000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107270050 Supplier Thermo Scientific Chemicals Supplier No. 107270050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 4870-65-9 | C8H7BrO2 | 215.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4870-65-9
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.04
InChI Key WAKFRZBXTKUFIW-UHFFFAOYSA-N
Synonym alpha-bromophenylacetic acid, bromo phenyl acetic acid, .alpha.-bromophenylacetic acid, bromophenylacetic acid, bromo-phenyl-acetic acid, bromo-phenylacetic acid, dl-.alpha.-bromophenylacetic acid, benzeneacetic acid, .alpha.-bromo, phenylbromoacetic acid, bromo phenyl aceticacid
PubChem CID 97919
IUPAC Name 2-bromo-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)Br

Specifications

Melting Point 82.0°C to 83.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH(Br)CO2H
Quantity 5 g
Beilstein 09,451
Solubility Information Solubility in water: insoluble
Formula Weight 215.04
Percent Purity 97%
Chemical Name or Material DL-α-Bromophenylacetic acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously wit
EINECSNumber 225-477-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.