Learn More
Medchemexpress LLC DL-Buthionine-S R-su 25mg | 25MG
SDP
Catalog No. 5000242260
Encompass_Preferred
DL-Buthionine-(S R)-sulfoximine hydrochloride (Buthionine sulfoximine hydrochloride) is a potent inhibitor of glutamylcysteine synthetase biosynthesis
Catalog No. 50-002-42260 Supplier Medchemexpress LLC Supplier No. HY106376B25MG
DL-Buthionine-(S R)-sulfoximine hydrochloride (Buthionine sulfoximine hydrochloride) is a potent inhibitor of glutamylcysteine synthetase biosynthesis