Learn More
Cayman Chemical DL-Carnitine-d9 (chloride), Purity: ≥99% deuterated forms (d1-d9), CAS: 1219386-75-0, 10 mg
SDP
Catalog No. 502935141
Encompass_Preferred
An internal standard for the quantification of carnitine by GC- or LC-MS
Catalog No. 50-293-5141 Supplier Cayman Chemical Supplier No. 2656610 MG
An internal standard for the quantification of carnitine by GC- or LC-MS