Learn More
Selleck Chemical LLC DL-Carnitine
SDP
Catalog No. 5000003661
Encompass_Preferred
DL-Carnitine (( )-Carnitin) is a racemic mixture of L-Carnitine and D-Carnitine that regulates fatty acid transport in the mitochondrion DL-carnitine is found to be ineffective or toxic for example to muscle cells and to the myocardium DL-Carnitine causes symptoms of myasthenia and cardiac arrhythmias
Catalog No. 50-000-03661 Supplier Selleck Chemical LLC Supplier No. S5937100MG
DL-Carnitine (( )-Carnitin) is a racemic mixture of L-Carnitine and D-Carnitine that regulates fatty acid transport in the mitochondrion DL-carnitine is found to be ineffective or toxic for example to muscle cells and to the myocardium DL-Carnitine causes symptoms of myasthenia and cardiac arrhythmias