Learn More
Medchemexpress LLC DL-carnitine-d9 chloride | 1219386-75-0 | MFCD03428232 | 98.0% | 206.72 | C7H7D9ClNO3 | 10 MG
SDP
Catalog No. 5000296251
Encompass_Preferred
(±)-Carnitine-d9 chloride is a deuterium-labeled racemic (DL) form of carnitine chloride used as an internal standard and tracer in quantitative analytical methods including LC-MS, GC-MS, and NMR. It is provided as a white to off-white solid with high assay and isotopic purity and is intended for research applications in fatty acid metabolism and pharmacokinetic studies.
  • Deuterium-labeled tracer for quantitative analysis.
  • Compatible with LC-MS, GC-MS, and NMR methods.
  • Racemic DL form suitable for general analytical use.
  • High assay and isotopic purity for reproducible results.
  • Supplied as a stable solid with defined cold storage recommendations.
Catalog No. 50-002-96251 Supplier Medchemexpress LLC Supplier No. HYB1453S110MG
(±)-Carnitine-d9 chloride is a deuterium-labeled racemic (DL) form of carnitine chloride used as an internal standard and tracer in quantitative analytical methods including LC-MS, GC-MS, and NMR. It is provided as a white to off-white solid with high assay and isotopic purity and is intended for research applications in fatty acid metabolism and pharmacokinetic studies.
  • Deuterium-labeled tracer for quantitative analysis.
  • Compatible with LC-MS, GC-MS, and NMR methods.
  • Racemic DL form suitable for general analytical use.
  • High assay and isotopic purity for reproducible results.
  • Supplied as a stable solid with defined cold storage recommendations.